Merck
  • Development and validation of a stability-indicating LC-UV method for the determination of pantethine and its degradation product based on a forced degradation study.
Development and validation of a stability-indicating LC-UV method for the determination of pantethine and its degradation product based on a forced degradation study.
Journal of pharmaceutical and biomedical analysis (2014-05-28)
Rossana Canavesi, Silvio Aprile, Elena Varese, Giorgio Grosa
ABSTRACT
Pantethine (d-bis-(N-pantothenyl-β-aminoethyl)-disulfide, PAN), the stable disulfide form of pantetheine, has beneficial effects in vascular diseases being able to decrease the hyperlipidaemia, moderate the platelet function and prevent the lipid peroxidation. Furthermore, recent studies suggested that PAN may be an effective therapeutic agent for cerebral malaria and, possibly, for neurodegenerative processes. Interestingly, in the literature, there were no data dealing with the chemical stability and the analytical aspects of PAN. Hence, in the present work the chemical stability of PAN was for the first time established through a forced degradation study followed by liquid chromatography tandem mass spectrometry investigation showing the formation of three degradation products of PAN (PD1, PD2 and POx) arising from hydrolytic, thermal and oxidative stresses. Based on these data a stability-indicating LC-UV method for simultaneous estimation of PAN, and its most relevant degradation product (PD1) was developed and validated; moreover the method allowed also the separation and the quantification of the preservative system, constituted by a paraben mixture. The method showed linearity for PAN (0.4-1.2mgmL(-1)), MHB, PHB (0.4-1.2μgmL(-1)) and PD1 (2.5-100μgmL(-1)); the precision, determined in terms of intra-day and inter-day precision, expressed as RSDs, were in the ranges 0.4-1.2 and 0.7-1.4, respectively. The method demonstrated to be accurate and robust; indeed the average recoveries were 100.2, 99.9, and 100.0% for PAN, MHB and PHB, respectively, and 99.9% for PD1. By applying small variations of the mobile phase composition, counter-ion concentration and pH the separation of analytes was not affected. Finally, the applicability of this method was evaluated analyzing the available commercial forms at release as well as during stability studies.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Supelco
Ethylene glycol, analytical standard
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Supelco
Methanol, analytical standard
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium pentanesulfonate, BioXtra
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium pentanesulfonate, ≥95% (elemental analysis)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
USP
Propylparaben, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Copper(II) sulfate pentahydrate, purum p.a., crystallized, ≥99.0% (RT)
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%