Merck
  • Polycyclic aromatic hydrocarbons associated with total suspended particles and surface soils in Kunming, China: distribution, possible sources, and cancer risks.
Polycyclic aromatic hydrocarbons associated with total suspended particles and surface soils in Kunming, China: distribution, possible sources, and cancer risks.
Environmental science and pollution research international (2014-11-26)
Xiaoxia Yang, Dong Ren, Wenwen Sun, Xiaoman Li, Bin Huang, Rong Chen, Chan Lin, Xuejun Pan
ABSTRACT
The concentrations, distribution, possible sources, and cancer risks of polycyclic aromatic hydrocarbons (PAHs) in total suspended particles (TSPs) and surface soils collected from the same sampling spots were compared in Kunming, China. The total PAH concentrations were 9.35-75.01 ng/m(3) and 101.64-693.30 ng/g dry weight (d.w.), respectively, in TSPs and surface soils. Fluoranthene (FLA), pyrene (PYR), chrysene (CHR), and phenanthrene (PHE) were the abundant compounds in TSP samples, and phenanthrene (PHE), fluorene (FLO), fluoranthene (FLA), benzo[b]fluoranthene (BbF), and benzo[g,h,i]perylene (BghiP) were the abundant compounds in surface soil samples. The spatial distribution of PAHs in TSPs is closely related to the surrounding environment, which varied significantly as a result of variations in source emission and changes in meteorology. However, the spatial distribution of PAHs in surface soils is supposed to correlate with a city's urbanization history, and high levels of PAHs were always observed in industry district, or central or old district of city. Based on the diagnostic ratios and principal component analysis (PCA), vehicle emissions (especially diesel-powered vehicles) and coal and wood combustion were the main sources of PAHs in TSPs, and the combustion of wood and coal, and spills of unburnt petroleum were the main sources of PAHs in the surface soils. The benzo[a]pyrene equivalent concentration (BaPeq) for the TSPs and surface soil samples were 0.16-2.57 ng/m(3) and 11.44-116.03 ng/g d.w., respectively. The incremental lifetime cancer risk (ILCR) exposed to particulate PAHs ranged from 10(-4) to 10(-3) indicating high potential of carcinogenic risk, and the ILCR exposed to soil PAHs was from 10(-7) to 10(-6) indicating virtual safety. These presented results showed that particle-bound PAHs had higher potential carcinogenic ability for human than soil PAHs. And, the values of cancer risk for children were always higher than for adults, which demonstrated that children were sensitive to carcinogenic effects of PAHs.
Product Description
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Supelco
Fluorene, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Perylene-d12, 98 atom % D
Sigma-Aldrich
Fluorene, 98%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Phenanthrene-d10, 98 atom % D, 98% (CP)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%