Merck
  • IL-17-mediated antifungal defense in the oral mucosa is independent of neutrophils.
IL-17-mediated antifungal defense in the oral mucosa is independent of neutrophils.
Mucosal immunology (2014-07-10)
K Trautwein-Weidner, A Gladiator, S Nur, P Diethelm, S LeibundGut-Landmann
ABSTRACT
Interleukin-17 (IL-17)-mediated immunity has emerged as a crucial host defense mechanism against Candida albicans infections in mucosal tissues and the skin. The precise mechanism by which the IL-17 pathway prevents fungal outgrowth has not been clarified. Neutrophils are critical for limiting fungal dissemination and IL-17 is generally thought to act by regulating neutrophil mobilization and trafficking to the site of infection. Using a mouse model of oropharyngeal candidiasis (OPC), we found that strikingly the IL-17 pathway is not required for the neutrophil response to C. albicans. Mice deficient for the IL-17 receptor subunits IL-17 receptor A (IL-17RA) or IL-17RC or mice depleted of IL-17A and IL-17F exhibited a normal granulocyte colony-stimulating factor (G-CSF) and CXC-chemokine response and displayed no defect in neutrophil recruitment or function. Instead, the inability of these mice to clear the fungus was associated with a selective defect in the induction of antimicrobial peptides (AMPs) in the epithelium that resulted in persistent fungal colonization. Importantly, this antifungal mechanism of IL-17A and IL-17F did not extend to the closely related family member IL-17C. Together, these data uncouple IL-17-dependent effector mechanisms from the neutrophil response and reveal a compartmentalization of the antifungal defense in the oral mucosa providing a new understanding of IL-17-mediated mucosal immunity against C. albicans.
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder