Merck
  • The antibiofilm activity of lingonberry flavonoids against oral pathogens is a case connected to residual complexity.
The antibiofilm activity of lingonberry flavonoids against oral pathogens is a case connected to residual complexity.
Fitoterapia (2014-06-01)
Kaisu R Riihinen, Zhen M Ou, Tanja Gödecke, David C Lankin, Guido F Pauli, Christine D Wu
ABSTRACT
The antimicrobial activity of lingonberry (Vaccinium vitis-idaea L.) was evaluated against two oral pathogens, Streptococcus mutans and Fusobacterium nucleatum. Long-bed gel permeation chromatography (GPC; Sephadex LH-20) yielded purified flavonoids, with the most efficient minimum inhibitory concentrations (MICs) against planktonic cells in the anthocyanin and procyanidin primary fractions against F. nucleatum (63-125 μg/ml) and in the procyanidin rich fraction against S. mutans (16-31 μg/ml). The purified flavonol glycosides and procyanidins inhibited biofilm formation of S. mutans (MICs 16-31 μg/ml), while the corresponding reference compounds showed no activity. Secondary GPC purification yielded flavonol glycosides devoid of antibiofilm activity in the 50% MeOH fraction, while elution with 70% acetone recovered a brownish material with activity against S. mutans biofilm (MIC 8 μg/ml). Even after HPLC-PDA, NMR, and MALDI-TOF analyses, the structural identity of this material remained unknown, while its color and analytical characteristics appear to be consistent with flavonoid oxidation products.
Product Description
Hyperoside, primary reference standard
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Supelco
Procyanidin B2, analytical standard
Sigma-Aldrich
Idaein chloride, ≥90% (HPLC)
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
USP
Quercetin, United States Pharmacopeia (USP) Reference Standard
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Chlorogenic acid, primary reference standard
Quercitrin, primary reference standard
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Quercetin 3-D-galactoside, ≥97.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, 99%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
trans-p-Coumaric acid, analytical standard
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)