Merck
  • Identification of transmembrane domain 1 & 2 residues that contribute to the formation of the ligand-binding pocket of the urotensin-II receptor.
Identification of transmembrane domain 1 & 2 residues that contribute to the formation of the ligand-binding pocket of the urotensin-II receptor.
Biochemical pharmacology (2014-09-02)
Xavier Sainsily, Jérôme Cabana, Brian J Holleran, Emanuel Escher, Pierre Lavigne, Richard Leduc
ABSTRACT
The vasoactive urotensin-II (UII), a cyclic undecapeptide widely distributed in cardiovascular, renal and endocrine systems, specifically binds the UII receptor (UT receptor), a G protein-coupled receptor (GPCR). The involvement of this receptor in numerous pathophysiological conditions including atherosclerosis, heart failure, hypertension, renal impairment and diabetes potentially makes it an interesting therapeutic target. To elucidate how UII binds the UT receptor through the identification of specific residues in transmembrane domains (TM) one (TM1) and two (TM2) that are involved in the formation of the receptor's binding pocket, we used the substituted-cysteine accessibility method (SCAM). Each residue of TM1 (V49((1.30)) to M76((1.57))) and TM2 (V88((2.41)) to H117((2.70))) was mutated, one by one, to a cysteine. The resulting mutants were then expressed in COS-7 cells and subsequently treated with the sulfhydryl-specific alkylating agent methanethiosulfonate-ethylammonium (MTSEA). MTSEA treatment resulted in a significant binding inhibition of (125)I-UII to mutant I54C((1.35)) in TM1 and mutants Y100C((2.53)), S103C((2.56)), F106C((2.59)), I107C((2.60)), T110C((2.63)) and Y111C((2.64)) in TM2. These results identify key structural residues in TM1 and TM2 that participate in the formation of the UT receptor binding pocket. Together with previous SCAM analysis of TM3, TM4, TM5, TM6 and TM7, these results have led us to identify residues within all 7 TMs that participate in UT's binding pocket and have enabled us to propose a model of this receptor's orthosteric binding site.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Terbium, foil, 50x50mm, thickness 0.1mm, as rolled, 99%
Terbium, rod, 50mm, diameter 6.35mm, cast, 99%
Terbium, foil, 25x25mm, thickness 0.1mm, as rolled, 99%
Terbium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Terbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Terbium, powder, 1g, max. particle size 500 micron, 99.9%
Terbium, powder, 2g, max. particle size 500 micron, 99.9%
Terbium, powder, 5g, max. particle size 500 micron, 99.9%
Terbium, rod, 100mm, diameter 6.35mm, cast, 99%