Merck
  • Using lysine-reactive fluorescent dye for surface characterization of a mAb.
Using lysine-reactive fluorescent dye for surface characterization of a mAb.
Journal of pharmaceutical sciences (2014-12-30)
Ming Lei, Yung-Hsiang Kao, Christian Schöneich
ABSTRACT
The biopharmaceutical industry increasingly demands thorough characterization of protein conformation and conformational dynamics to ensure product quality and consistency. Here, we present a chromatography-based method that is able to characterize protein conformation and conformational dynamics at peptide level resolution in a high-throughput manner. The surface lysine residues of the protein were labeled with a fluorescent dye prior to enzyme digestion. The resulting peptide maps were monitored by fluorescence detection where fluorescence peak area indicates higher solvent accessibility at a specific site. The peptides of reactivity difference and the extent of the difference can be detected by HPLC with fluorescent detector alone, whereas the identity of these peptides can then be determined by mass spectrometry if desired. We first demonstrated this method is suitable for probing protein surface/conformation by studying the effect of deglycosylation on a recombinant mAb, IgG 1. We then applied our method to study the interaction of the mAb with a common excipient, polysorbate-20 (PS-20). The presence of PS-20 increased the fluorescent labeling of several lysine residues on the mAb. This result provides a first insight into PS20-mAb interaction at peptide level resolution.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium iodoacetate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Sodium iodoacetate, ≥98%, BioUltra
Sigma-Aldrich
Sodium iodoacetate, ≥98%
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C