Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Pricing and availability is not currently available.
Sigma-Aldrich
Rifabutin, >98% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Azithromycin for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Azithromycin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Azithromycin, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
USP
Azithromycin Identity, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Supelco
Metronidazole, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur™, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Pricing and availability is not currently available.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Metronidazole, BioXtra
Pricing and availability is not currently available.
Supelco
Metronidazole, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Pricing and availability is not currently available.