Merck
  • Position and length of fatty acids strongly affect receptor selectivity pattern of human pancreatic polypeptide analogues.
Position and length of fatty acids strongly affect receptor selectivity pattern of human pancreatic polypeptide analogues.
ChemMedChem (2014-08-27)
Veronika Mäde, Kathrin Bellmann-Sickert, Anette Kaiser, Jens Meiler, Annette G Beck-Sickinger
ABSTRACT
Pancreatic polypeptide (PP) is a satiety-inducing gut hormone targeting predominantly the Y4 receptor within the neuropeptide Y multiligand/multireceptor family. Palmitoylated PP-based ligands have already been reported to exert prolonged satiety-inducing effects in animal models. Here, we suggest that other lipidation sites and different fatty acid chain lengths may affect receptor selectivity and metabolic stability. Activity tests revealed significantly enhanced potency of long fatty acid conjugates on all four Y receptors with a preference of position 22 over 30 at Y1 , Y2 and Y5 receptors. Improved Y receptor selectivity was observed for two short fatty acid analogues. Moreover, [K(30)(E-Prop)]hPP2-36 (15) displayed enhanced stability in blood plasma and liver homogenates. Thus, short chain lipidation of hPP at key residue 30 is a promising approach for anti-obesity therapy because of maintained selectivity and a sixfold increased plasma half-life.
Sigma-Aldrich
Bromotrimethylsilane, purum, ≥97.0% (AT)
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bromotrimethylsilane, 97%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥99%
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
HATU, ≥98.0% (CHN)
Sigma-Aldrich
Thioanisole, ReagentPlus®, ≥99%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
HATU, 97%
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%