Merck
  • Determination of perfluorinated alkyl acids in corn, popcorn and popcorn bags before and after cooking by focused ultrasound solid-liquid extraction, liquid chromatography and quadrupole-time of flight mass spectrometry.
Determination of perfluorinated alkyl acids in corn, popcorn and popcorn bags before and after cooking by focused ultrasound solid-liquid extraction, liquid chromatography and quadrupole-time of flight mass spectrometry.
Journal of chromatography. A (2014-07-06)
Cristina Moreta, María Teresa Tena
ABSTRACT
An analytical method is proposed to determine ten perfluorinated alkyl acids (PFAAs) [nine perfluorocarboxylic acids (PFCAs) and perfluorooctane sulfonate (PFOS)] in corn, popcorn and microwave popcorn packaging by focused ultrasound solid-liquid extraction (FUSLE) and ultra high performance liquid chromatography (UHPLC) coupled to quadrupole-time of flight mass spectrometry (QTOF-MS/MS). Selected PFAAs were extracted efficiently in only one 10-s cycle by FUSLE, a simple, safe and inexpensive technique. The developed method was validated for microwave popcorn bags matrix as well as corn and popcorn matrices in terms of linearity, matrix effect error, detection and quantification limits, repeatability and recovery values. The method showed good accuracy with recovery values around 100% except for the lowest chain length PFAAs, satisfactory reproducibility with RSDs under 16%, and sensitivity with limits of detection in the order of hundreds picograms per gram of sample (between 0.2 and 0.7ng/g). This method was also applied to the analysis of six microwave popcorn bags and the popcorn inside before and after cooking. PFCAs contents between 3.50ng/g and 750ng/g were found in bags, being PFHxA (perfluorohexanoic acid) the most abundant of them. However, no PFAAs were detected either corn or popcorn, therefore no migration was assumed.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%
Sigma-Aldrich
Perfluorooctanoic acid, 95%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Perfluorodecanoic acid, 98%