Merck
  • Two-phase hollow fiber liquid phase microextraction based on magnetofluid for simultaneous determination of Echinacoside, Tubuloside B, Acteoside and Isoacteoside in rat plasma after oral administration of Cistanche salsa extract by high performance liquid chromatography.
Two-phase hollow fiber liquid phase microextraction based on magnetofluid for simultaneous determination of Echinacoside, Tubuloside B, Acteoside and Isoacteoside in rat plasma after oral administration of Cistanche salsa extract by high performance liquid chromatography.
Journal of pharmaceutical and biomedical analysis (2014-02-18)
Jun Zhou, Qiong Zhang, Jiang Bing Sun, Xiao Li Sun, Ping Zeng
ABSTRACT
A new and fast sample preparation technique based on two-phase hollow fiber liquid phase microextraction (HF-LPME) with magnetofluid was developed to quantitate and determine the four phenylethanoid glycosides (PhGs) (Echinacoside, Tubuloside B, Acteoside and Isoacteoside) in plasma after oral administration of Cistanche salsa extract. Analysis was accomplished by reversed-phase high performance liquid chromatography (RP-HPLC) with ultraviolet detection. Parameters that affect the HF-LPME processes, such as the content of magnetic powder, the solvent type, salt content, stirring speed, extraction time and hollow fiber length, were investigated and optimized. Under the optimized conditions, the preconcentration factors for PhGs were higher than 625. The calibration curve for PhGs was linear in the range of 0.1-100ngmL(-1) with correlation coefficients greater than 0.9996. The intra-day and inter-day precision (RSD) were below 8.74% and the limits of detection (LOD) for the four PhGs were 8-15pgmL(-1) (S/N=3). The validated method was successfully applied to separate and determine the four PhGs in rat plasma after oral administration of C. salsa extract.
Product Description
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Decanol, 98%
Sigma-Aldrich
1-Decanol, ≥98%
Supelco
Dodecane, analytical standard
Supelco
Toluene, analytical standard
Supelco
Ethyl acetate, analytical standard
Supelco
Hexane, analytical standard
Supelco
Density Standard 749 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
1-Decanol, analytical standard
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Hexane, anhydrous, 95%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material