Merck
  • Identification of a DNA methylation signature in blood cells from persons with Down Syndrome.
Identification of a DNA methylation signature in blood cells from persons with Down Syndrome.
Aging (2015-02-24)
Maria Giulia Bacalini, Davide Gentilini, Alessio Boattini, Enrico Giampieri, Chiara Pirazzini, Cristina Giuliani, Elisa Fontanesi, Maria Scurti, Daniel Remondini, Miriam Capri, Guido Cocchi, Alessandro Ghezzo, Alberto Del Rio, Donata Luiselli, Giovanni Vitale, Daniela Mari, Gastone Castellani, Mario Fraga, Anna Maria Di Blasio, Stefano Salvioli, Claudio Franceschi, Paolo Garagnani
ABSTRACT
Down Syndrome (DS) is characterized by a wide spectrum of clinical signs, which include segmental premature aging of central nervous and immune systems. Although it is well established that the causative defect of DS is the trisomy of chromosome 21, the molecular bases of its phenotype are still largely unknown. We used the Infinium HumanMethylation450 BeadChip to investigate DNA methylation patterns in whole blood from 29 DS persons, using their relatives (mothers and unaffected siblings) as controls. This family-based model allowed us to monitor possible confounding effects on DNA methylation patterns deriving from genetic and environmental factors. Although differentially methylated regions (DMRs) displayed a genome-wide distribution, they were enriched on chromosome 21. DMRs mapped in genes involved in developmental functions, including embryonic development (HOXA family) and haematological (RUNX1 and EBF4) and neuronal (NCAM1) development. Moreover, genes involved in the regulation of chromatin structure (PRMD8, KDM2B, TET1) showed altered methylation. The data also showed that several pathways are affected in DS, including PI3K-Akt signaling. In conclusion, we identified an epigenetic signature of DS that sustains a link between developmental defects and disease phenotype, including segmental premature aging.
Product Description
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Thioctic acid containing impurity B, European Pharmacopoeia (EP) Reference Standard
Thioctic acid, European Pharmacopoeia (EP) Reference Standard
Thioctic acid for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Alpha Lipoic Acid, United States Pharmacopeia (USP) Reference Standard
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
(±)-α-Lipoic acid, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
(±)-α-Lipoic acid, synthetic, ≥99% (titration), powder
Sigma-Aldrich
(±)-α-Lipoic acid, ≥98.0%
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%