Merck
  • Deficient sucrose synthase activity in developing wood does not specifically affect cellulose biosynthesis, but causes an overall decrease in cell wall polymers.
Deficient sucrose synthase activity in developing wood does not specifically affect cellulose biosynthesis, but causes an overall decrease in cell wall polymers.
The New phytologist (2014-06-13)
Lorenz Gerber, Bo Zhang, Melissa Roach, Umut Rende, András Gorzsás, Manoj Kumar, Ingo Burgert, Totte Niittylä, Björn Sundberg
ABSTRACT
The biosynthesis of wood in aspen (Populus) depends on the metabolism of sucrose, which is the main transported form of carbon from source tissues. The largest fraction of the wood biomass is cellulose, which is synthesized from UDP-glucose. Sucrose synthase (SUS) has been proposed previously to interact directly with cellulose synthase complexes and specifically supply UDP-glucose for cellulose biosynthesis. To investigate the role of SUS in wood biosynthesis, we characterized transgenic lines of hybrid aspen with strongly reduced SUS activity in developing wood. No dramatic growth phenotypes in glasshouse-grown trees were observed, but chemical fingerprinting with pyrolysis-GC/MS, together with micromechanical analysis, showed notable changes in chemistry and ultrastructure of the wood in the transgenic lines. Wet chemical analysis showed that the dry weight percentage composition of wood polymers was not changed significantly. However, a decrease in wood density was observed and, consequently, the content of lignin, hemicellulose and cellulose was decreased per wood volume. The decrease in density was explained by a looser structure of fibre cell walls as shown by increased wall shrinkage on drying. The results show that SUS is not essential for cellulose biosynthesis, but plays a role in defining the total carbon incorporation to wood cell walls.
Product Description
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
Galactose, European Pharmacopoeia (EP) Reference Standard
Millipore
D-(−)-Arabinose, suitable for microbiology, ≥99.0%
Xylose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Sigma-Aldrich
D-(−)-Arabinose, ≥98% (GC)
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-Glucuronic acid, ≥98% (GC)
Sigma-Aldrich
L-(−)-Fucose, ≥98% (GC)
Supelco
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Galactose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG