Merck
  • Porphyromonas gulae 41-kDa fimbriae induced osteoclast differentiation and cytokine production.
Porphyromonas gulae 41-kDa fimbriae induced osteoclast differentiation and cytokine production.
The Journal of veterinary medical science (2014-11-26)
Haruka Sasaki, Kiyoko Watanabe, Toshizo Toyama, Yasunori Koyata, Nobushiro Hamada
ABSTRACT
Porphyromonas gulae is considered to be associated with canine periodontitis. We have previously reported that the P. gulae American Type Culture Collection (ATCC) 51700 comprised 41-kDa fimbriae. The purpose of the present study was to demonstrate the roles of 41-kDa fimbrial protein in periodontal disease. In this study, we examined the involvement of the 41-kDa fimbrial protein in osteoclast differentiation and cytokine production in murine macrophages. Furthermore, alveolar bone resorption induced by P. gulae infection in rats was evaluated. To estimate osteoclast differentiation, bone marrow cells and MC3T3-G2/PA6 cells were cultured with or without the 41-kDa fimbrial protein for 7 days. BALB/c mouse peritoneal macrophages were stimulated with the 41-kDa fimbrial protein, and the levels of interleukin (IL)-1β and tumor necrosis factor (TNF)-α production were determined by enzyme-linked immunosorbent assay. Osteoclast differentiation was significantly enhanced by treatment with the 41-kDa fimbrial protein in a dose-dependent manner. The total area of pits formed on the dentine slices with osteoclasts incubated with the 41-kDa fimbrial protein was significantly greater than that of the control. The purified 41-kDa fimbrial protein induced IL-1β and TNF-α production in BALB/c mouse peritoneal macrophages after 6 hr of incubation in a dose-dependent manner. The bone loss level in rats infected with P. gulae was significantly higher than that of the sham-infected rats. These results suggest that P. gulae 41-kDa fimbriae play important roles in the pathogenesis of periodontal disease.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)