Merck
  • Formulation strategy and evaluation of nanocrystal piroxicam orally disintegrating tablets manufacturing by freeze-drying.
Formulation strategy and evaluation of nanocrystal piroxicam orally disintegrating tablets manufacturing by freeze-drying.
International journal of pharmaceutics (2014-04-01)
Francesco Lai, Elena Pini, Francesco Corrias, Jacopo Perricci, Maria Manconi, Anna Maria Fadda, Chiara Sinico
ABSTRACT
Piroxicam (PRX) is a non-steroidal anti-inflammatory drug characterized by a poor water solubility and consequently by a low oral bioavailability. In this work, different nanocrystal orally disintegrating tablets (ODT) were prepared to enhance piroxicam dissolution rate and saturation solubility. PRX nanocrystals were prepared by means of high pressure homogenization technique using poloxamer 188 as stabilizer. Three different ODTs were prepared with the same nanosuspension using different excipients in order to study their effect on the PRX dissolution properties. PRX nanocrystal size and zeta potential were determined by photon correlation spectroscopy. Additional characterization of PRX nanocrystal ODT was carried out by infrared spectroscopy, X-ray powder diffractometry, differential scanning calorimetry. Dissolution study was performed in distilled water (pH 5.5) and compared with PRX coarse suspension ODT, PRX/poloxamer 188 physical mixture, bulk PRX samples and a PRX commercial ODT. All PRX nanocrystal ODT formulations showed a higher drug dissolution rate than coarse PRX ODT. PRX nanocrystal ODT prepared using gelatin or croscarmellose as excipient showed a higher PRX dissolution rate compared with the commercial formulation and ODT prepared using xanthan gum. Overall results confirmed that improved PRX dissolution rate is due to the increased surface-to-volume ratio due to the nanosized drug particle but also revealed the important role of different excipients used.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)