Merck
  • Control of Maillard-type off-flavor development in ultrahigh-temperature-processed bovine milk by phenolic chemistry.
Control of Maillard-type off-flavor development in ultrahigh-temperature-processed bovine milk by phenolic chemistry.
Journal of agricultural and food chemistry (2014-07-30)
Smaro Kokkinidou, Devin G Peterson
ABSTRACT
The application of phenolic compounds to suppress Maillard chemistry and off-flavor development in ultrahigh-termperature (UHT)-processed milk during processing and storage was investigated. Five phenolic compounds were examined for structure-reactivity relationships (catechin, genistein, daidzein, 1,2,3-trihydroxybenzene, and 1,3,5-trihydroxybenzene). The levels of key transient Maillard reaction (MR) intermediates (reactive carbonyl species) and select off-flavor markers (methional, 2-acetyl-2-thiazoline, 2-acetyl-1-pyrroline) were quantified by LC-MS/MS and GC-MS/ToF, respectively. The addition of phenolic compounds prior to UHT processing significantly reduced the concentration of MR intermediates and related off-flavor compounds compared to a control sample (p < 0.05). All phenolic compounds demonstrated unique structure reactivity and, notably, those with a more activated A-ring for aromatic electrophilic substitution (catechin, genistein, and 1,3,5-trihydroxybenzene) showed the strongest suppression effect on the off-flavor markers and reactive carbonyl species. Sensory studies were in agreement with the analytical data. The cooked flavor intensity was rated lower for the recombination model samples of the catechin-treated UHT milk compared to the control UHT milk. Additionally, consumer acceptability studies showed catechin-treated UHT milk to have significantly higher liking scores when compared the control sample (Fisher's LSD = 0.728).
Sigma-Aldrich
Pyrogallol, ACS reagent
Supelco
2,3-Butanedione, analytical standard
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
2,3-Butanedione, 97%
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Methional, ≥97%, FG
Sigma-Aldrich
Hydroxyacetone, contains ≤500 ppm sodium carbonate as stabilizer, technical grade, 90%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Methional, natural, ≥98%, FG
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Pyrogallol, analytical standard
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.
Sigma-Aldrich
Phloroglucinol, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Supelco
(+)-Catechin, analytical standard
Sigma-Aldrich
3-(Methylthio)propionaldehyde, 96%
Phloroglucinol (anhydrous), European Pharmacopoeia (EP) Reference Standard