Skip to Content
Merck
All Photos(1)

Documents

Y0000493

Phloroglucinol (anhydrous)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Phloroglucinol, 1,3,5-Trihydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
1341907
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phloroglucinol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1cc(O)cc(O)c1

InChI

1S/C6H6O3/c7-4-1-5(8)3-6(9)2-4/h1-3,7-9H

InChI key

QCDYQQDYXPDABM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phloroglucinol (anhydrous) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Inder Pal Singh et al.
Expert opinion on therapeutic patents, 19(6), 847-866 (2009-05-29)
Phloroglucinol compounds, both synthetic as well as natural, have shown a vast array of biological activities. There are a wide range of applications of phloroglucinol compounds in pharmaceuticals, cosmetics, textiles, paints and dyeing industries. Although many of the phloroglucinols have
Fang Yang et al.
Applied microbiology and biotechnology, 93(2), 487-495 (2011-11-22)
Phloroglucinol derivatives are a major class of secondary metabolites of wide occurrence in biological systems. In the bacteria kingdom, these compounds can only be synthesized by some species of Pseudomonads. Pseudomonas spp. could produce 2,4-diacetylphloroglucinol (DAPG) that plays an important
Phloroglucinol compounds of natural origin.
Inder Pal Singh et al.
Natural product reports, 23(4), 558-591 (2006-07-29)
Inder Pal Singh et al.
Natural product reports, 27(3), 393-416 (2010-02-25)
This review covers the synthetic aspects of various naturally occurring phloroglucinols, describing syntheses of 42 compounds and citing 137 references.
Ya-Qiang He et al.
Chinese medical journal, 128(7), 956-962 (2015-04-04)
Phloroglucinol plays an important role in oxidative stress and inflammatory responses. The effects of phloroglucinol have been proven in various disease models. The aim of the present study was to investigate the efficacy and possible mechanisms of phloroglucinol in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service