Merck
  • Binding mechanisms for histamine and agmatine ligands in plasmid deoxyribonucleic acid purifications.
Binding mechanisms for histamine and agmatine ligands in plasmid deoxyribonucleic acid purifications.
Journal of chromatography. A (2014-09-30)
Ângela Sousa, Patrícia Pereira, Fani Sousa, João A Queiroz
ABSTRACT
Histamine and agmatine amino acid derivatives were immobilized into monolithic disks, in order to combine the specificity and selectivity of the ligand with the high mass transfer and binding capacity offered by monolithic supports, to purify potential plasmid DNA biopharmaceuticals. Different elution strategies were explored by changing the type and salt concentration, as well as the pH, in order to understand the retention pattern of different plasmids isoforms The pVAX1-LacZ supercoiled isoform was isolated from a mixture of pDNA isoforms by using NaCl increasing stepwise gradient and also by ammonium sulfate decreasing stepwise gradient, in both histamine and agmatine monoliths. Acidic pH in the binding buffer mainly strengthened ionic interactions with both ligands in the presence of sodium chloride. Otherwise, for histamine ligand, pH values higher than 7 intensified hydrophobic interactions in the presence of ammonium sulfate. In addition, circular dichroism spectroscopy studies revealed that the binding and elution chromatographic conditions, such as the combination of high ionic strength with extreme pH values can reversibly influence the structural stability of the target nucleic acid. Therefore, ascending sodium chloride gradients with pH manipulation can be preferable chromatographic conditions to be explored in the purification of plasmid DNA biopharmaceuticals, in order to avoid the environmental impact of ammonium sulfate.
Product Description
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Pricing and availability is not currently available.
Sigma-Aldrich
Histamine dihydrochloride, ≥99% (TLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Histamine dihydrochloride, ≥99.0% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Pricing and availability is not currently available.
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
SAFC
Tromethamine
Pricing and availability is not currently available.
Supelco
Ampicillin, analytical standard
Pricing and availability is not currently available.
Histamine dihydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Agmatine sulfate salt, ≥97%, powder
Pricing and availability is not currently available.