Merck
  • Resveratrol loaded gelatin nanoparticles synergistically inhibits cell cycle progression and constitutive NF-kappaB activation, and induces apoptosis in non-small cell lung cancer cells.
Resveratrol loaded gelatin nanoparticles synergistically inhibits cell cycle progression and constitutive NF-kappaB activation, and induces apoptosis in non-small cell lung cancer cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-03-18)
Subburayan Karthikeyan, Sugeerappa Laxmanappa Hoti, Nagarajan Rajendra Prasad
ABSTRACT
Previously, we reported that the prepared resveratrol (RSV) loaded gelatin nanoparticles (GNPs) possessed enhanced anticancer effect than free RSV in non-small cell lung carcinoma cells and Swiss albino mice. The present study aims to explore the relevant mechanism of cell death induced by the combination of RSV-GNPs in NCI-H460 cells. To increase its bioavailability and anticancer efficacy, we have encapsulated RSV-GNPs by Coacervation method. The detailed methods of preparation and characterization of RSV-GNPs were reported in our earlier publication. RSV-GNPs treated cells showed a further increased level of lipid peroxidative markers, i.e. TBARS and LHP in NCI-H460 cells. Activities of antioxidant enzymes SOD, CAT, GPx and GSH levels were decreased upon the treatment with RSV-GNPs in NCI-H460 cells. The nuclear fragmentation was evaluated by DAPI staining and data showed condensed apoptotic bodies upon treatment with the combination of RSV-GNPs compared to RSV alone treatment group. In addition, cell death induced by RSV-GNPs was mainly due to apoptosis which was characterized by a nuclear DNA fragmentation in a ladder-pattern was obtained from the genomic DNA analysis. Moreover, Western blotting analysis showed that apoptosis induced by RSV-GNPs is associated with the increased Bax, p53, p21, caspase-3 protein levels, and decreased Bcl-2 and NF-κB proteins expression, which indicates the involvement of mitochondria-dependent apoptosis in the anticancer efficacy of RSV-GNPs in NCI-H460 cells. It was also found that this enhanced anticancer efficacy of RSV-GNPs induced cell arrest in the G0/G1 phase of cell cycle. Taken together, the results of our study clearly suggested that the cell death induced by the combination of RSV-GNPs would involve alteration in expression of p53, p21, caspase-3, Bax, Bcl-2 and NF-κB, indicating oxidative mechanism in NCI-H460 cells. Based on these results, it is concluded that GNPs is an ideal way to deliver RSV because of its high loading efficiency and superior efficacy in NCI-H460 cells.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Supelco
Acetone, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard