Merck
  • Highly magneto-responsive multilayer microcapsules for controlled release of insulin.
Highly magneto-responsive multilayer microcapsules for controlled release of insulin.
International journal of pharmaceutics (2014-08-28)
Chunli Zheng, Yafei Ding, Xiaoqing Liu, Yunkai Wu, Liang Ge
ABSTRACT
In this study, magneto-responsive polyelectrolyte multilayer microcapsules were successfully prepared by the formation of shell with biocompatible iron oxide nanoparticles (Fe₃O₄ NPs) and polyallylamine hydrochloride (PAH) by layer-by-layer (LbL) self-assembly technique. The self-assembled microcapsules were characterized by SEM, TEM and zeta-potential analyzer. According to the pH sensitivity of the microcapsule membrane permeability, insulin was encapsulated, with the encapsulation efficiency of 92.08±5.57%. The in vitro release behavior in an external alternating magnetic field indicated that once the magnetic field was applied, the drug release was greatly accelerated. In addition, according to the observed pulse release upon cyclic on-off operations of magnetic field, it could be assumed that the magneto-responsive microcapsules had an excellent "switching on" effect, which might be attributed to the rearrangement of shell structure caused by magnetic nanoparticles twisting and polyelectrolyte chains shaking, hence the increase of microcapsule membrane permeability and the enhancement of insulin release.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
2-Propen-1-amine hydrochloride, AldrichCPR
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)