Merck
  • Identification of TRIML2, a novel p53 target, that enhances p53 SUMOylation and regulates the transactivation of proapoptotic genes.
Identification of TRIML2, a novel p53 target, that enhances p53 SUMOylation and regulates the transactivation of proapoptotic genes.
Molecular cancer research : MCR (2014-09-27)
Che-Pei Kung, Sakina Khaku, Matthew Jennis, Yan Zhou, Maureen E Murphy
ABSTRACT
The tumor-suppressor protein p53, encoded by TP53, inhibits tumorigenesis by inducing cell-cycle arrest, senescence, and apoptosis. Several genetic polymorphisms exist in TP53, including a proline to arginine variant at amino acid 72 (P72 and R72, respectively); this polymorphism alters p53 function. In general, the P72 variant shows increased ability to induce cell-cycle arrest, whereas the R72 variant possesses increased ability to induce apoptosis, relative to P72. At present, the underlying mechanisms for these functional differences are not fully understood. Toward elucidating the molecular basis for these differences, a gene-expression microarray analysis was conducted on normal human fibroblast cells that are homozygous for P72 and R72 variants, along with subclones of these lines that express a p53 short hairpin (shp53). Approximately three dozen genes were identified whose transactivation is affected by the codon 72 polymorphism. One of these is the tripartite-motif family-like 2 (TRIML2) gene, which is preferentially induced by the R72 variant. Importantly, the accumulated data indicate that TRIML2 interacts with p53, and facilitates the modification of p53 with SUMO2. TRIML2 also enhances the ability of p53 to transactivate a subset of proapoptotic target genes associated with prolonged oxidative stress, including PIDD, PIG3 (TP53I3), and PIG6 (PRODH). These data indicate that TRIML2 is part of a feed-forward loop that activates p53 in cells expressing the R72 variant, particularly after prolonged stress. The defined actions of TRIML2, in part, explain the underlying molecular basis for increased apoptotic potential of the R72 variant of p53.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Supelco
Ginkgolic acid C15:1, analytical standard
Sigma-Aldrich
Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1), liquid, clone DO-1, Calbiochem®
Supelco
5-Fluorouracil, analytical standard
SAFC
BIS-TRIS
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
SAFC
BIS-TRIS
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Etoposide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder