Merck
  • Assessment of non-steroidal anti-inflammatory and analgesic pharmaceuticals in seawaters of North of Portugal: occurrence and environmental risk.
Assessment of non-steroidal anti-inflammatory and analgesic pharmaceuticals in seawaters of North of Portugal: occurrence and environmental risk.
The Science of the total environment (2014-12-08)
Aleksandar Lolić, Paula Paíga, Lúcia H M L M Santos, Sandra Ramos, Manuela Correia, Cristina Delerue-Matos
ABSTRACT
The occurrence of seven pharmaceuticals and two metabolites belonging to non-steroidal anti-inflammatory drugs and analgesics therapeutic classes was studied in seawaters. A total of 101 samples covering fourteen beaches and five cities were evaluated in order to assess the spatial distribution of pharmaceuticals among north Portuguese coast. Seawaters were selected in order to embrace different bathing water quality (excellent, good and sufficient). Acetaminophen, ketoprofen and the metabolite hydroxyibuprofen were detected in all the seawater samples at maximum concentrations of 584, 89.7 and 287 ng L(-1), respectively. Carboxyibuprofen had the highest seawater concentration (1227 ng L(-1)). The temporal distribution of the selected pharmaceuticals during the bathing season showed that, in general, higher concentrations were detected in August and September. The environmental risk posed by the pharmaceuticals detected in seawaters towards different trophic levels (fish, daphnids and algae) was also assessed. Only diclofenac showed hazard quotients above one for fish, representing a potential risk for aquatic organisms. These results were observed in seawaters classified as excellent bathing water. Additional data is needed in order to support the identification and prioritization of risks posed by pharmaceuticals in marine environment.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Hydroxyibuprofen, VETRANAL®, analytical standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Ketoprofen, meets USP testing specifications
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
Nimesulide
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Ketoprofen, ≥98% (TLC)
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetaminophen, analytical standard
Supelco
Nimesulide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ketoprofen, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
USP
Ketoprofen, United States Pharmacopeia (USP) Reference Standard
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Naproxen, European Pharmacopoeia (EP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard