Merck
  • TMEM16A is a Ca(2+) -activated Cl(-) channel expressed in the renal collecting duct.
TMEM16A is a Ca(2+) -activated Cl(-) channel expressed in the renal collecting duct.
Acta physiologica (Oxford, England) (2014-06-11)
P Svenningsen, M R Nielsen, N Marcussen, S Walter, B L Jensen
ABSTRACT
In the renal collecting ducts, ATP stimulates a Ca(2+) -activated chloride current. The identity of the channel responsible for the current under physiological conditions is not known and it was hypothesized that TMEM16a is a relevant candidate in the renal collecting duct. The cortical collecting duct cell line M-1 was used as a model of the collecting duct. The ATP induced Ca(2+) signalling was imaged in cells loaded with Ca(2+) -sensitive fluorescent probes using confocal laser-scanning fluorescence microscopy. Chloride current was determined by mounting M-1 cell layers in Ussing chamber. The expression of TMEM16a in human kidney was tested by immunohistochemistry. M-1 cells displayed a transient increase in intracellular Ca(2+) concentration in response to 100 nm ATP. This response was completely blocked by addition of 100 μm suramin, indicating that ATP signals through purinergic P2 receptors. Apical addition of 100 nm ATP induced a Cl(-) current, which was blocked by suramin, DPC and the cysteine-modifying compound MTSET. M-1 cells were found to express TMEM16a at the mRNA and protein level. Functionally, it was found that knock-down of TMEM16a expression in M-1 cells inhibited the ATP induced Cl(-) -current. In human and mouse kidney sections, TMEM16a protein expression was localized to the collecting duct, and TMEM16a was found to be excreted in human urinary exosomes. TMEM16a is a Ca(2+) -activated Cl(-) channel expressed in the collecting ducts.
Sigma-Aldrich
Imidazole, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
1,2,4,5-Tetramethylbenzene, 98%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tyms
Sigma-Aldrich
MISSION® esiRNA, targeting human TYMS
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Imidazole, ≥99% (titration), free-flowing, Redi-Dri
Supelco
1,2,4,5-Tetramethylbenzene, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting human TBXAS1
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
DAPI, for nucleic acid staining