Merck
  • Directed targeting of chromatin to the nuclear lamina is mediated by chromatin state and A-type lamins.
Directed targeting of chromatin to the nuclear lamina is mediated by chromatin state and A-type lamins.
The Journal of cell biology (2015-01-07)
Jennifer C Harr, Teresa Romeo Luperchio, Xianrong Wong, Erez Cohen, Sarah J Wheelan, Karen L Reddy
ABSTRACT
Nuclear organization has been implicated in regulating gene activity. Recently, large developmentally regulated regions of the genome dynamically associated with the nuclear lamina have been identified. However, little is known about how these lamina-associated domains (LADs) are directed to the nuclear lamina. We use our tagged chromosomal insertion site system to identify small sequences from borders of fibroblast-specific variable LADs that are sufficient to target these ectopic sites to the nuclear periphery. We identify YY1 (Ying-Yang1) binding sites as enriched in relocating sequences. Knockdown of YY1 or lamin A/C, but not lamin A, led to a loss of lamina association. In addition, targeted recruitment of YY1 proteins facilitated ectopic LAD formation dependent on histone H3 lysine 27 trimethylation and histone H3 lysine di- and trimethylation. Our results also reveal that endogenous loci appear to be dependent on lamin A/C, YY1, H3K27me3, and H3K9me2/3 for maintenance of lamina-proximal positioning.
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
1 Estimated to ship on March 07, 2025
$167.00
100 g
1 Estimated to ship on March 07, 2025
$261.00
500 g
1 Estimated to ship on March 07, 2025
$997.00
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
L-Lysine, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
1 Estimated to ship on March 07, 2025
$92.70
Lysine acetate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Estimated to ship on March 07, 2025
$88.80
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
1 Available to ship on January 06, 2025
Details...
$177.00
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
1 Estimated to ship on March 07, 2025
$126.00