Skip to Content
Merck
All Photos(1)

Key Documents

67448

Supelco

L-Lysine monohydrochloride

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(S)-2,6-Diaminohexanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65
Beilstein:
3563889
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

mp

263 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC([C@@H](N)CCCCN)=O.[H]Cl

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1

InChI key

BVHLGVCQOALMSV-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Lysine monohydrochloride is generally used as a food and feed supplement.It can also find applications in medicine and pharmaceutical industries.L-Lysine monohydrochloride can be produced via sugar fermentation by modified microorganisms.[1]
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intratec Solutions
L-Lysine-HCl Production from Glucose - Cost Analysis - Lysine E12A (2016)
Lubin Jiang et al.
Nature, 499(7457), 223-227 (2013-07-05)
The variant antigen Plasmodium falciparum erythrocyte membrane protein 1 (PfEMP1), which is expressed on the surface of P. falciparum-infected red blood cells, is a critical virulence factor for malaria. Each parasite has 60 antigenically distinct var genes that each code
Nicholas I Proellocks et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(7), 3103-3113 (2014-04-08)
The genomes of malaria parasites (Plasmodium spp.) contain a family of genes encoding proteins with a Plasmodium helical interspersed subtelomeric (PHIST) domain, most of which are predicted to be exported into the parasite-infected human red blood cell (iRBC). Here, using
Guntur Fibriansah et al.
Nature communications, 6, 6341-6341 (2015-02-24)
Dengue virus (DENV) infects ~400 million people annually. There is no licensed vaccine or therapeutic drug. Only a small fraction of the total DENV-specific antibodies in a naturally occurring dengue infection consists of highly neutralizing antibodies. Here we show that
Lunzhi Dai et al.
Nature chemical biology, 10(5), 365-370 (2014-04-01)
We report the identification of a new type of histone mark, lysine 2-hydroxyisobutyrylation (Khib), and identify the mark at 63 human and mouse histone Khib sites, including 27 unique lysine sites that are not known to be modified by lysine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service