Merck
  • The active form of vitamin D, calcitriol, induces a complex dual upregulation of endothelin and nitric oxide in cultured endothelial cells.
The active form of vitamin D, calcitriol, induces a complex dual upregulation of endothelin and nitric oxide in cultured endothelial cells.
American journal of physiology. Endocrinology and metabolism (2014-10-23)
Patricia Martínez-Miguel, Jose Manuel Valdivielso, Diana Medrano-Andrés, Pablo Román-García, Jose Luis Cano-Peñalver, Manuel Rodríguez-Puyol, Diego Rodríguez-Puyol, Susana López-Ongil
ABSTRACT
Despite the presence of vitamin D receptor (VDR) in endothelial cells, the effect of vitamin D on endothelial function is unknown. An unbalanced production of vasoactive endothelial factors such as nitric oxide (NO) or endothelin-1 (ET-1) results in endothelial dysfunction, which can alter the normal cardiovascular function. Present experiments were devoted to assess the effect of active vitamin D (calcitriol) on the synthesis of endothelial vasoactive factors. The results were that, in cells, calcitriol increased ET-1 and NO productions, which were measured by ELISA and fluorimetric assay, respectively. Calcitriol also increased endothelin-converting enzyme-1 (ECE-1) and endothelial-nitric oxide synthase (eNOS) activities, their mRNA (qPCR), their protein expressions (Western-blot), and their promoter activities (transfection assays). Calcitriol did not change prepro-ET-1 mRNA. The effect was specific to VDR activation because when VDR was silenced by siRNA, the observed effects disappeared. Mechanisms involved in each upregulation differed. ECE-1 upregulation depended on AP-1 activation, whereas eNOS upregulation depended directly on VDR activation. To evaluate the in vivo consequences of acute calcitriol treatment, normal Wistar rats were treated with a single ip injection of 400 ng/kg calcitriol and euthanized 24 h later. Results confirmed those observed in cells, that production and expression of both factors were increased by calcitriol. Besides, calcitriol-treated rats showed a slight rise in mean blood pressure, which decreased when pretreated with FR-901533, an ECE-1 antagonist. We conclude that calcitriol increases the synthesis of both ET-1 and NO in endothelial cells. However, the ET-1 upregulation seems to be biologically more relevant, as animals acutely treated with calcitriol show slight increases in blood pressure.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)