Merck
  • In vitro activities of amphotericin B deoxycholate and liposomal amphotericin B against 604 clinical yeast isolates.
In vitro activities of amphotericin B deoxycholate and liposomal amphotericin B against 604 clinical yeast isolates.
Journal of medical microbiology (2014-09-12)
Maria Teresa Montagna, Grazia Lovero, Caterina Coretti, Osvalda De Giglio, Domenico Martinelli, Andrea Bedini, Mario Delia, Antonio Rosato, Mauro Codeluppi, Giuseppina Caggiano
ABSTRACT
We determined the in vitro antifungal activity of liposomal amphotericin B (L-AmB) against 604 clinical yeast isolates. Amphotericin B deoxycholate (D-AmB) was tested in parallel against all the isolates. Susceptibility testing was performed according to the Clinical and Laboratory Standards Institute (CLSI) M27-A3 method. Overall, L-AmB was highly active against the isolates (mean MIC, 0.42 µg ml(-1); MIC90, 1 µg ml(-1); 97.2 % of MICs were ≤1 µg ml(-1)) and comparable to D-AmB (mean MIC, 0.48 µg ml(-1); MIC90, 1 µg ml(-1); 97.3 % of MICs were ≤1 µg ml(-1)). The in vitro activity of D-AmB and L-AmB was correlated (R(2) = 0.61; exp(b), 2.3; 95 % CI, 2.19-2.44, P<0.001). Candida albicans (mean MICs of D-AmB and L-AmB, 0.39 µg ml(-1) and 0.31 µg ml(-1), respectively) and Candida parapsilosis (mean MICs of D-AmB and L-AmB, 0.38 µg ml(-1) and 0.35 µg ml(-1), respectively) were the species most susceptible to the agents tested, while Candida krusei (currently named Issatchenkia orientalis) (mean MICs of D-AmB and L-AmB, 1.27 µg ml(-1) and 1.13 µg ml(-1), respectively) was the least susceptible. The excellent in vitro activity of L-AmB may have important implications for empirical treatment approaches and support its role in treatment of a wide range of invasive infections due to yeasts.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Supelco
D-(+)-Glucose, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Pricing and availability is not currently available.
Amphotericin B for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Pricing and availability is not currently available.
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
SAFC
Sodium deoxycholate
Pricing and availability is not currently available.