Merck
  • Evaluation of seasonal and daily changes of plasma thyroxine and cortisol levels in wild masu salmon Oncorhynchus masou, sampled by a Japanese fishing method.
Evaluation of seasonal and daily changes of plasma thyroxine and cortisol levels in wild masu salmon Oncorhynchus masou, sampled by a Japanese fishing method.
Journal of fish biology (2014-09-30)
A Munakata, G Miura, H Matsuda
ABSTRACT
A new fish sampling method was developed using a Japanese bait fishing rod (8-9 m carbon rod and a nylon line with a small fine wire single hook), which is considered to catch wild salmonid juveniles with low sampling stress. Using this method, seasonal and daily changes of plasma thyroxine (T4 ) and cortisol levels were examined in wild parr, pre-smolts and smolts of masu salmon Oncorhynchus masou in contiguous locations in a coastal river (Kesen River; 44 km) in northern Honshu Island, Japan, overlapping the period of smoltification and seaward migration from August to March. Plasma T4 and cortisol were low in 0+ and 1+ year parr caught in August and September. In March, some yearling (1+ year) fish, which were judged as pre-smolts, and smolts appeared mainly in mid and lower reaches, while parr (0+ and 1+ year parr) continued to appear in the upper and mid reaches. In March, 1+ year pre-smolts and smolts showed high plasma T4 levels while the levels of 1+ year parr were low. During March 2008-2010, plasma T4 levels of 1+ year pre-smolts and smolts had high levels from early to mid-March, whereas plasma cortisol levels of 1+ year smolts were low in early March and increased towards mid-March. Based on these data, plasma cortisol increases probably occur following the increases of plasma T4 levels to lead the 1+ year O. masou to the completion of smoltification and initiation of seaward migration.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard