- Sensitivity of Danio rerio (Teleostei, Cyprinidae) during two stages of development based on acute toxicity tests.
Sensitivity of Danio rerio (Teleostei, Cyprinidae) during two stages of development based on acute toxicity tests.
Bulletin of environmental contamination and toxicology (2014-08-30)
R Freiry, J A A Stelzer, L Maltchik, A Arenzon
PMID25168694
ABSTRACT
The sensitivity of Danio rerio to three chemicals was compared at two growth stages [larval (10 ± 2 after hatching) and post-larval (60 ± 4 days after hatching)] based on acute toxicity tests. Thirty-nine 48 h acute toxicity tests were performed with the substances CuSO4, NaCl and KCl. The 48 h LC50 values at the two growth stages were compared by independent samples t-tests. The results showed a clear decrease in sensitivity when post-larval organisms were used. Since acute toxicity test methods for D. rerio that recommend using post-larval stage fish do not represent the most sensitive stage of the test organism, our study suggests a revision of the methods to use larval fish.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%