Merck
  • Resolvin D2 supports MCF-7 cell proliferation via activation of estrogen receptor.
Resolvin D2 supports MCF-7 cell proliferation via activation of estrogen receptor.
The Journal of pharmacology and experimental therapeutics (2014-08-01)
Nuha Al-Zaubai, Cameron N Johnstone, May May Leong, John Li, Mark Rizzacasa, Alastair G Stewart
ABSTRACT
Inflammation has been implicated in tumor initiation, angiogenesis, and metastasis, and linked to the development of more aggressive, therapy-resistant estrogen receptor (ER)-positive breast cancer. Resolvin D2 (RvD2) is a potent anti-inflammatory lipid mediator. As RvD2 may be synthesized within breast tumors by both tumor cells and the surrounding stroma cells and is present in plasma at bioactive concentrations, we sought to characterize the impact of RvD2 on cell processes underlying breast tumor growth and spread. Trypan-blue exclusion, transfection with estrogen response element (ERE) reporter, real-time quantitative polymerase chain reaction, competitive radioligand binding assays, Western blotting, and immunofluorescence were the techniques used. Unexpectedly, whereas RvD2 (10-1000 nM) supported the proliferation of the ER-positive breast tumor (MCF-7) cells, it did not affect the ER-negative MDA-MB-231 cell number. The proliferative effect of RvD2 in MCF-7 cells was attenuated by the ER antagonist ICI 182,780 (7α-[9-[(4,4,5,5,5-pentafluoropentyl)sulfinyl]nonyl]estra-1,3,5(10)-triene-3,17β-diol). Furthermore, RvD2 increased ERE transcriptional activity in a number of ER-positive breast and ovarian tumor cell lines. This activation was also inhibited by ICI 182,780. RvD2 altered the expression of a subset of estrogen-responsive genes. Although binding experiments showed that RvD2 did not directly compete with [(3)H]17β-estradiol for ER binding, prior exposure of MCF-7 cells to RvD2 resulted in a significant reduction in the apparent cytosolic ER density. Confocal immunocytochemistry and Western blotting studies showed that RvD2 promoted nuclear localization of ERα. These observations indicate that RvD2 displays significant but indirect estrogenic properties and has the potential to play a role in estrogen-dependent breast cancer progression.
Product Description
Supelco
cis-4,7,10,13,16,19-Docosahexaenoic acid, analytical standard
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture