Merck
  • Determination of traces of copper and zinc in honeys by the solid phase extraction pre-concentration followed by the flame atomic absorption spectrometry detection.
Determination of traces of copper and zinc in honeys by the solid phase extraction pre-concentration followed by the flame atomic absorption spectrometry detection.
Environmental monitoring and assessment (2014-05-30)
Helena Stecka, Dominika Jedryczko, Maja Welna, Pawel Pohl
ABSTRACT
A simple and fast solid phase extraction procedure was developed to pre-concentrate traces of Cu and Zn prior to their determination in honey samples by flame atomic absorption spectrometry. The sample preparation included dissolution of honey samples and the passage (at 20 ml/min) of resulting 10% m/v solutions (100 ml) through Dowex 50W × 8-400 resin beds in order to quantitatively retain Cu and Zn and separate them from the glucose and fructose matrix. Enriched Cu and Zn traces were recovered with 5.0 ml of a 3.0 mol/l HCl solution and quantified by flame atomic absorption spectrometry. The procedure proposed was used to analyze sixty nine commercially available and freshly ripened honey samples coming from the Lower Silesia region (Poland). It enabled to measure Cu and Zn within the range of 0.01-1.42 and 0.03-15.38 μg/g, respectively, with precision better than 4%. Accuracy, assessed on the basis of the recovery test and the comparison of results with those obtained using wet digestion and inductively coupled plasma optical emission spectrometry, was ranged from -4% to +6%. Detection limits of Cu and Zn achieved with this method were 5 and 7 ng/g, respectively.
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Copper - O.F.H.C., tube, 1000mm, outside diameter 0.38mm, inside diameter 0.22mm, wall thickness 0.08mm, 99.95+%
Copper - O.F.H.C., rod, 500mm, diameter 9.5mm, as drawn, 99.95+%
Copper - O.F.H.C., wire reel, 20m, diameter 0.125mm, annealed, 99.95+%
Copper - O.F.H.C., rod, 100mm, diameter 76mm, as drawn, 99.95+%
Copper - O.F.H.C., wire reel, 2m, diameter 1.5mm, hard, 99.95+%
Copper - O.F.H.C., tube, 100mm, outside diameter 3.2mm, inside diameter 2.4mm, wall thickness 0.4mm, 99.95+%