Merck
Rat atrial responses to Bothrops jararacussu (jararacuçu) snake venom.
Toxicology (2014-06-29)
Mariana A P Rodrigues, Lourdes Dias, André L Rennó, Norma C Sousa, Adriana Smaal, Delano A da Silva, Stephen Hyslop
ABSTRACT
Envenoming by the pitviper Bothrops jararacussu produces cardiovascular alterations, including coagulopathy, systemic hemorrhage, hypotension, circulatory shock and renal failure. In this work, we examined the activity of this venom in rat isolated right atria. Incubation with venom (0.025, 0.05, 0.1 and 0.2mg/ml) caused concentration-dependent muscle contracture that was not reversed by washing. Muscle damage was seen histologically and confirmed by quantification of creatine kinase-MB (CK-MB) release. Heating and preincubation of venom with p-bromophenacyl bromide (a phospholipase A2 inhibitor) abolished the venom-induced contracture and muscle damage. In contrast, indomethacin, a non-selective inhibitor of cyclooxygenase, and verapamil, a voltage-gated Ca(2+) channel blocker, did not affect the responses to venom. Preincubation of venom with Bothrops or Bothrops/Crotalus antivenom or the addition of antivenom soon after venom attenuated the venom-induced changes in atrial function and tissue damage. These results indicate that B. jararacussu venom adversely affected rat atrial contractile activity and muscle organization through the action of venom PLA2; these venom-induced alterations were attenuated by antivenom.
Sigma-Aldrich
Creatine Phosphokinase from bovine heart, Type III, salt-free, lyophilized powder, ≥30 units/mg protein
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,4′-Dibromoacetophenone, >98%
Supelco
2,4′-Dibromoacetophenone, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)