Merck
  • Diversity-oriented synthesis of azapeptides with basic amino acid residues: aza-lysine, aza-ornithine, and aza-arginine.
Diversity-oriented synthesis of azapeptides with basic amino acid residues: aza-lysine, aza-ornithine, and aza-arginine.
Organic letters (2014-06-25)
Mariam Traoré, Ngoc-Duc Doan, William D Lubell
ABSTRACT
Aza-peptides with basic amino acid residues (lysine, ornithine, arginine) and derivatives were synthesized by an effective approach featuring alkylation of a hydrazone-protected aza-glycine residue with α-bromo ω-chloro propane and butane to provide the corresponding alkyl chloride side chains. Displacement of the chloride with azide and various amines gave entry to azaOrn, azaLys, and azaArg containing peptides as demonstrated by the solution and solid-phase syntheses of 29 examples, including an aza-library of Growth Hormone Releasing Peptide-6 analogs.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
SAFC
L-Arginine
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
DL-Ornithine monohydrochloride, ≥99.0% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
[D-Trp7, Ala8, D-Phe10]-α-Melanocyte Stimulating Hormone Amide Fragment 6-11, ≥97% (HPLC)
Arginine, European Pharmacopoeia (EP) Reference Standard
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-Ornithine monohydrochloride, ≥99%
Sigma-Aldrich
L-Ornithine monohydrochloride, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
L-Ornithine monohydrochloride, BioXtra, ≥99%
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
D-Ornithine monohydrochloride, ~98%
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
L-Ornithine dihydrochloride, ≥99.0% (AT)
Sigma-Aldrich
L-Ornithine hydrochloride, 99%
Lysine acetate, European Pharmacopoeia (EP) Reference Standard