Merck
Structures of acylated sucroses from the flower buds of Prunus mume.
Journal of natural medicines (2014-02-18)
Katsuyoshi Fujimoto, Seikou Nakamura, Takahiro Matsumoto, Tomoe Ohta, Masayuki Yoshikawa, Keiko Ogawa, Eri Kashiwazaki, Hisashi Matsuda
ABSTRACT
Seven new acylated sucroses, mumeoses P-V, were isolated from the flower buds of Prunus mume, cultivated in Zhejiang province, China. Their chemical structures were elucidated on the basis of chemical and physicochemical evidence. Moreover, mumeoses C, D, and R, were shown to substantially inhibit aldose reductase.
Product Description
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard