Merck
  • Verapamil, and its metabolite norverapamil, inhibit macrophage-induced, bacterial efflux pump-mediated tolerance to multiple anti-tubercular drugs.
Verapamil, and its metabolite norverapamil, inhibit macrophage-induced, bacterial efflux pump-mediated tolerance to multiple anti-tubercular drugs.
The Journal of infectious diseases (2014-02-18)
Kristin N Adams, John D Szumowski, Lalita Ramakrishnan
ABSTRACT
Drug tolerance likely represents an important barrier to tuberculosis treatment shortening. We previously implicated the Mycobacterium tuberculosis efflux pump Rv1258c as mediating macrophage-induced tolerance to rifampicin and intracellular growth. In this study, we infected the human macrophage-like cell line THP-1 with drug-sensitive and drug-resistant M. tuberculosis strains and found that tolerance developed to most antituberculosis drugs, including the newer agents moxifloxacin, PA-824, linezolid, and bedaquiline. Multiple efflux pump inhibitors in clinical use for other indications reversed tolerance to isoniazid and rifampicin and slowed intracellular growth. Moreover, verapamil reduced tolerance to bedaquiline and moxifloxacin. Verapamil's R isomer and its metabolite norverapamil have substantially less calcium channel blocking activity yet were similarly active as verapamil at inhibiting macrophage-induced drug tolerance. Our finding that verapamil inhibits intracellular M. tuberculosis growth and tolerance suggests its potential for treatment shortening. Norverapamil, R-verapamil, and potentially other derivatives present attractive alternatives that may have improved tolerability.
Sigma-Aldrich
Piperine, ≥95%, FG
Sigma-Aldrich
Piperine, ≥97%
Sigma-Aldrich
D-Cycloserine
Sigma-Aldrich
4-Aminosalicylic acid, 99%
Sigma-Aldrich
Clofazimine
Sigma-Aldrich
D-Cycloserine, synthetic
Sigma-Aldrich
D-Cycloserine
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Rifampicin, European Pharmacopoeia (EP) Reference Standard
Supelco
Piperine, analytical standard
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Piperine, European Pharmacopoeia (EP) Reference Standard
Clofazimine, European Pharmacopoeia (EP) Reference Standard
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ethionamide for system suitability, European Pharmacopoeia (EP) Reference Standard
Ethionamide, European Pharmacopoeia (EP) Reference Standard
Rifabutin, European Pharmacopoeia (EP) Reference Standard
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Rifabutin, >98% (HPLC), powder
Sigma-Aldrich
(±)-Norverapamil hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Ethionamide
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
USP
Rifabutin, United States Pharmacopeia (USP) Reference Standard