Skip to Content
Merck
All Photos(1)

Key Documents

1603800

USP

Rifabutin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Ansamycin, Ansatipine (Farmitalia), LM-427, Mycobutin (Farmitalia)

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$544.00

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$544.00

About This Item

Empirical Formula (Hill Notation):
C46H62N4O11
CAS Number:
Molecular Weight:
847.00
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$544.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

rifabutin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[N+H]1(CCC2(NC3=C4NC(=O)\C(=C/C=C/[C@@H]([C@@H]([C@H]([C@H]([C@H]([C@@H]([C@@H]([C@H](\C=C\O[C@]5(Oc6c(c(c(c(c6C)[O-])C4=O)C3=N2)C5=O)C)OC)C)OC(=O)C)C)O)C)O)C)\C)CC1)CC(C)C

InChI

1S/C46H62N4O11/c1-22(2)21-50-18-16-46(17-19-50)48-34-31-32-39(54)28(8)42-33(31)43(56)45(10,61-42)59-20-15-30(58-11)25(5)41(60-29(9)51)27(7)38(53)26(6)37(52)23(3)13-12-14-24(4)44(57)47-36(40(32)55)35(34)49-46/h12-15,20,22-23,25-27,30,37-38,41,49,52-54H,16-19,21H2,1-11H3,(H,47,57)/b13-12+,20-15+,24-14-/t23-,25+,26+,27+,30-,37-,38+,41+,45-/m0/s1

InChI key

ATEBXHFBFRCZMA-VXTBVIBXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rifabutin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Rifabutin Capsules
  • Rifabutin Compounded Oral Suspension

Biochem/physiol Actions

Rifabutin is an antibiotic; antitumor.
Rifabutin is an antibiotic; antitumor. Rifabutin interferes with HSP-90 molecular chaperone, enhances ubiquitination and protein degradation, and inactivates bacterial RNA polymerase.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Against the proposition: all patients with advanced HIV disease should be offered rifabutin prophylaxis.
A Pozniak et al.
Genitourinary medicine, 72(4), 269-271 (1996-08-01)
Timothy J Haggerty et al.
PloS one, 9(12), e114506-e114506 (2014-12-17)
In an effort to enhance antigen-specific T cell recognition of cancer cells, we have examined numerous modulators of antigen-expression. In this report we demonstrate that twelve different Hsp90 inhibitors (iHsp90) share the ability to increase the expression of differentiation antigens
Anne M Baciewicz et al.
Current medical research and opinion, 29(1), 1-12 (2012-11-10)
Rifampin is a potent inducer of both cytochrome P-450 oxidative enzymes and the P-glycoprotein transport system. Among numerous well documented, clinically significant interactions, examples include warfarin, oral contraceptives, itraconazole, digoxin, verapamil, simvastatin, and human immunodeficiency virus-related protease inhibitors. Rifabutin reduces
A Loeliger et al.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease, 16(1), 6-15 (2011-08-09)
To assess how to best manage co-administration of rifabutin (RFB) and human immunodeficiency virus 1 (HIV-1) protease inhibitor (PI) containing antiretroviral treatment (ART). Recommended for initial anti-tuberculosis treatment, rifampicin (RMP) lowers PI concentrations below therapeutic levels, posing significant challenges for
J P Gisbert et al.
Alimentary pharmacology & therapeutics, 35(2), 209-221 (2011-12-02)
Even with the current most effective treatment regimens, a relevant proportion of patients will fail to eradicate Helicobacter pylori infection. To evaluate the role of rifabutin in the treatment of H. pylori infection. Bibliographical searches were performed in MEDLINE. Data

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service