Merck
  • Current challenges and concepts of the thermomechanical treatment of nickel-titanium instruments.
Current challenges and concepts of the thermomechanical treatment of nickel-titanium instruments.
Journal of endodontics (2013-01-17)
Ya Shen, Hui-min Zhou, Yu-feng Zheng, Bin Peng, Markus Haapasalo
ABSTRACT
The performance and mechanical properties of nickel-titanium (NiTi) instruments are influenced by factors such as cross-section, flute design, raw material, and manufacturing processes. Many improvements have been proposed by manufacturers during the past decade to provide clinicians with safer and more efficient instruments. The mechanical performance of NiTi alloys is sensitive to their microstructure and associated thermomechanical treatment history. Heat treatment or thermal processing is one of the most fundamental approaches toward adjusting the transition temperature in NiTi alloy, which affects the fatigue resistance of NiTi endodontic files. The newly developed NiTi instruments made from controlled memory wire, M-Wire (Dentsply Tulsa Dental Specialties, Tulsa, OK), or R-phase wire represent the next generation of NiTi alloys with improved flexibility and fatigue resistance. The advantages of NiTi files for canal cleaning and shaping are decreased canal transportation and ledging, a reduced risk of file fracture, and faster and more efficient instrumentation. The clinician must understand the nature of different NiTi raw materials and their impact on instrument performance because many new instruments are introduced on a regular basis. This review summarizes the metallurgical properties of next-generation NiTi instruments, the impact of thermomechanical treatment on instrument flexibility, and the resistance to cyclic fatigue and torsion. The aim of this review was to provide clinicians with the knowledge necessary for evidence-based practices, maximizing the benefits from the selection and application of NiTi rotary instruments for root canal treatment.
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Titanium, wire reel, 5m, diameter 0.75mm, as drawn, 99.6+%
Titanium, wire reel, 5m, diameter 1.0mm, as drawn, 99.6+%
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Nickel, rod, 100mm, diameter 12.0mm, 99%
Nickel, rod, 100mm, diameter 30mm, 99%
Nickel, foil, 2m coil, thickness 0.125mm, coil width 50mm, annealed, 99%
Nickel, wire reel, 10m, diameter 0.01mm, as drawn, 99.98%
Nickel, foil, 2m coil, thickness 0.50mm, annealed, 99%
Titanium, tube, 1000mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Nickel, rod, 100mm, diameter 10.0mm, 99.99+%
Nickel, foil, 0.5m coil, thickness 0.125mm, annealed, 99%
Nickel, tube, 500mm, outside diameter 1.8mm, inside diameter 1.6mm, wall thickness 0.1mm, hard, 99.5%
Nickel, wire reel, 100m, diameter 0.25mm, hard, 99.98%
Nickel, foil, 4mm disks, thickness 0.007mm, 99.95%
Titanium, tube, 1000mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Nickel, foil, 10mm disks, thickness 0.0125mm, 99.9%