Merck
Biodegradation of sulfamethoxazole: current knowledge and perspectives.
Applied microbiology and biotechnology (2012-08-18)
Simone Larcher, Viviane Yargeau
ABSTRACT
Antibiotic compounds, like sulfamethoxazole (SMX), have become a concern in the aquatic environment due to the potential development of antibacterial resistances. Due to extensive consumption, excretion and disposal, SMX has been frequently detected in wastewaters and surface waters. This has led to numerous studies investigating the nature of SMX, with many researchers focusing on the biodegradation and persistence of SMX during wastewater treatment and in the environment. This review provides a summary of recent developments, outlines the discrepancies in observations and results, and demonstrates the need for further research to determine optimal biological removal strategies for SMX and other antibiotics.
Product Description
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sulfamethoxazole
USP
Sulfamethoxazole, United States Pharmacopeia (USP) Reference Standard
Sulfamethoxazole, European Pharmacopoeia (EP) Reference Standard