Merck
  • Development of palladium-catalyzed transformations using propargylic compounds.
Development of palladium-catalyzed transformations using propargylic compounds.
Chemical & pharmaceutical bulletin (2012-03-03)
Masahiro Yoshida
ABSTRACT
It is known that propargylic compounds having an ester and a halide at the propargylic positions react with palladium complexes leading to π-propargylpalladium and allenylpalladium complexes, which cause various transformations in the presence of the reactants. The aim of the present study was to develop novel palladium-catalyzed transformations using propargylic compounds. As diastereoselective reactions of propargylic compounds with bis-nucleophiles, we have developed palladium-catalyzed reactions of propargylic carbonates with 2-substituted cyclohexane-1,3-diones, 2-(2-hydroxyphenyl)acetates and 2-oxocyclohex-3-enecarboxylates. These processes produce highly substituted cyclic compounds in a highly stereoselective manner. Through our studies on the construction of substituted 2,3-allenols by the reactions of propargylic oxiranes, it has been made clear that palladium-catalyzed coupling reactions occur in the presence of arylboronic acids and terminal alkynes. The processes can be carried out in mild conditions to yield substituted 4-aryl-2,3-allenols in a diastereoselective manner. In our attempt to develop CO2-recycling reactions, we developed a methodology for the synthesis of cyclic carbonates by palladium-catalyzed reactions of propargylic carbonates with phenols. Our findings suggested that the process proceeds through a pathway involving decarboxylation-followed fixation of the liberated CO2. Diastereoselective, enantioselective, and enantiospecific construction of cyclic carbonates have been achieved by the application of this methodology.
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Palladium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.75%
Palladium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, 8mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, light tested, 100x100mm, thickness 0.047mm, as rolled, 99.95%
Palladium, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.95%
Palladium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%
Palladium, wire reel, 0.5m, diameter 0.02mm, as drawn, 99.95%
Palladium, wire reel, 2m, diameter 0.25mm, as drawn, 99.99+%
Palladium, wire reel, 10m, diameter 0.25mm, hard, 99.95%
Palladium, rod, 100mm, diameter 2.0mm, 99.95%
Palladium, rod, 10mm, diameter 6.0mm, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.008mm, 99.95%
Palladium, wire reel, 1m, diameter 0.25mm, hard, 99.95%
Palladium, microfoil, disks, 25mm, thinness 0.05μm, specific density 63.6μg/cm2, permanent mylar 3.5μm support, 99.99%
Palladium, tube, 50mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%