Merck
  • Identification of differentiation-stage specific markers that define the ex vivo osteoblastic phenotype.
Identification of differentiation-stage specific markers that define the ex vivo osteoblastic phenotype.
Bone (2014-07-02)
Natalie A Twine, Li Chen, Chi N Pang, Marc R Wilkins, Moustapha Kassem
ABSTRACT
The phenotype of osteoblastic (OB) cells in culture is currently defined using a limited number of markers of low sensitivity and specificity. For the clinical use of human skeletal (stromal, mesenchymal) stem cells (hMSC) in therapy, there is also a need to identify a set of gene markers that predict in vivo bone forming capacity. Thus, we used RNA sequencing to examine changes in expression for a set of skeletally-related genes across 8 time points between 0 and 12days of ex vivo OB differentiation of hMSC. We identified 123 genes showing significant temporal expression change. Hierarchical clustering and Pearson's correlation generated 4 groups of genes: early stage differentiation genes (peak expression: 0-24h, n=28) which were enriched for extracellular matrix organisation, e.g. COL1A1, LOX, and SERPINH1; middle stage differentiating genes (peak expression days: 3 and 6, n=20) which were enriched for extracellular matrix/skeletal system development e.g. BMP4, CYP24A1, and TGFBR2; and late stage differentiation genes (peak expression days: 9 and 12, n=27) which were enriched for bone development/osteoblast differentiation, e.g. BMP2 and IGF2. In addition, we identified 13 genes with bimodal temporal expression (2 peaks of expression: days 0 and 12) including VEGFA, PDGFA and FGF2. We examined the specificity of the 123 genes' expression in skeletal tissues and thus propose a set of ex vivo differentiation-stage-specific markers (n=21). In an independent analysis, we identified a subset of genes (n=20, e.g. ELN, COL11A1, BMP4) to predict the bone forming capacity of hMSC and another set (n=20, e.g. IGF2, TGFB2, SMAD3) associated with the ex vivo phenotype of hMSC obtained from osteoporotic patients.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard