Merck
  • Engineering bamboo-type TiO2 nanotube arrays to enhance their photocatalytic property.
Engineering bamboo-type TiO2 nanotube arrays to enhance their photocatalytic property.
Journal of nanoscience and nanotechnology (2014-04-18)
Dongsheng Guan, Paul J Hymel, Chengjun Zhou, Ying Wang
ABSTRACT
Bamboo-type TiO2 nanotube arrays with high surface area can be synthesized by alternating voltage (AV) anodization for their important use as photocatalytic medium. Their morphologies are highly dependent on preparation parameters including anodization time and electrolyte composition. Minimum time of high-voltage steps required for forming desired bamboo ridge spacing on these nanotubes can be calculated from current-time profiles recorded during potentiostatic anodization at the voltage. Water content in NH4F-containing ethylene glycol (EG) electrolytes is optimized simply from analyses of current transients or current-voltage relations for anodization in EG electrolytes with different amount of water, in order to achieve efficient electrochemical growth of TiO2 nanotubes for large ridge density and long tube length. Two types of bamboo-type TiO2 nanotubes with the same length of 5.46 microm but different ridge spacing are synthesized for photocatalytic degradation of methylene blue (MB) under UV radiation. Both of the bamboo-type nanotube arrays show improved photo catalysis compared to smooth TiO2 nanotubes of the same length, due to their larger surface area favorable for heterogeneous catalytic processes. In particular, the apparent rate constant of photocatalytic degradation on bamboo-type nanotubes is up to 29.4% higher than that for degradation on smooth ones.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Titanium, rod, 200mm, diameter 2mm, as drawn, 99.99+%
Titanium, tube, 100mm, outside diameter 4.25mm, inside diameter 3.75mm, wall thickness 0.25mm, annealed, 99.6+%
Titanium, microfoil, disks, 10mm, thinness 1.0μm, specific density 429μg/cm2, permanent mylar 3.5μm support, 99.6+%
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Titanium, wire reel, 0.5m, diameter 1.0mm, annealed, 99.6+%
Titanium, wire reel, 25m, diameter 0.20mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 0.51mm, inside diameter 0.35mm, wall thickness 0.08mm, annealed, 99.6+%
Titanium, tube, 500mm, outside diameter 2.03mm, inside diameter 1.55mm, wall thickness 0.24mm, annealed, 99.6+%
Titanium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.6+%
Titanium, rod, 200mm, diameter 25mm, annealed, 99.6+%
Titanium, wire, straight, 1000mm, diameter 1.0mm, as drawn, 99.6+%
Titanium, wire reel, 1m, diameter 1.5mm, as drawn, 99.6+%
Titanium, wire reel, 2m, diameter 1.0mm, as drawn, 99.99+%
Titanium, wire reel, 5m, diameter 0.20mm, annealed, 99.6+%
Titanium, wire reel, 20m, diameter 0.125mm, annealed, 99.6+%
Titanium, wire reel, 20m, diameter 0.05mm, hard, 99.8%