- Enhanced photocatalytic efficacy of hetropolyacid pillared TiO2 nanocomposites.
Enhanced photocatalytic efficacy of hetropolyacid pillared TiO2 nanocomposites.
Journal of nanoscience and nanotechnology (2014-04-18)
R Nivea, V Gunasekaran, R Kannan, T Sakthivel, K Govindan
PMID24738400
ABSTRACT
The removal of dye from industrial effluents is prime important, photo-catalysis is a finest method to combat dye from effluents. This study concerns about the investigation of photocatalytic activity of TiO2-HPAs (Hetropolyacids) nanocomposite namely TiO2-Phosphomolybdic nanocomposite [TiO2-HMA] and TiO2-Phosphotungstic nanocomposite [TiO2-HWA] which were prepared by Sol-gel method and the same were characterized by using XRD, SEM-EDAX. The photocatalytic activity of prepared photo-catalysts were evaluated and compared by the degradation of Methylene Blue dye in water solution under UV irradiation. In that TiO2-HMA nanocomposite showed superior photocatalytic activity than TiO2-HWA.
Product Description
Titanium, rod, 100mm, diameter 20mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, rod, 100mm, diameter 3.0mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, mesh, 100x100mm, nominal aperture 4.3mm, wire diameter 1.5mm, 4.4x4.4 wires/inch, open area 94%, platinized diamond mesh
Titanium, sphere, 100pcs, diameter 3mm, 99.6+%
Titanium, tube, 200mm, outside diameter 6.1mm, inside diameter 5.1mm, wall thickness 0.5mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 3.0mm, inside diameter 1.5mm, wall thickness 0.75mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 50mm, diameter 8.0mm, as drawn, 99.99+%
Titanium, tube, 200mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, rod, 25mm, diameter 9.5mm, annealed, 99.99+%
Titanium, tube, 200mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis