Merck
Effect of TiO2 on the photocatalytic properties of bismuth oxide.
Environmental technology (2014-04-08)
Gourav Singla, K Singh
ABSTRACT
Bi2-xTixO3+x/2 (x = 0.05, 0.10 and 0.15) photocatalysts are prepared by solid-state technique. The band gap of the prepared sample was estimated from the onset of UV-Vis absorption spectra. The photocatalytic activities of as prepared samples are investigated under UV light irradiation for the decomposition of methyl orange (MO). It has been observed that the rate of decomposition of MO increased with increasing Ti4+ content in the present samples. Moreover, it shows better photocatalytic activity than undoped Bi2O3 and TiO2. The lowest band gap is observed for x = 0.15 sample, i.e. 2.55 eV, which also showed the highest photocatalytic activity in the present sample.
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Methyl Orange, for microscopy (Hist.), indicator (pH 3.0-4.4)
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Methyl Orange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, powder, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, ReagentPlus®, powder, 10 μm, 99.9% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, nanopowder, 90-210 nm particle size, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Bismuth(III) oxide, purum, ≥98.0% (KT)
Bismuth, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 150x150mm, thickness 0.02mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 50x50mm, thickness 0.0025mm, permanent polyester support, 99.97%
Bismuth, foil, light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%