Merck
  • Photodegradation of dioxin in contaminated soil in the presence of solvents and nanoscale TiO2 particles.
Photodegradation of dioxin in contaminated soil in the presence of solvents and nanoscale TiO2 particles.
Environmental technology (2014-04-08)
Nguyen Duy Binh, Nguyen Thi Kim Oanh, Preeda Parkpian
ABSTRACT
Decomposition of 2,3,7,8-Tetrachlorodibenzo-p-dioxin (2,3,7,8-TCDD) present in soil under ultraviolet (UV) illumination (350-400nm) was investigated using a combination of nontoxic solvents mixed in soil and nanoscale anatase TiO2 (nTiO2) distributed on 2mm top soil surface. Three types of UV-exposure experiments were conducted: intermittent exposure (8 h/day) for 90 days and 120 days, sequential intermittent (120 days) and continuous (24 h/day) for the next 55 days, and continuous exposure for 55 days. The influence of several factors on dioxin photodegradation efficiency was investigated, including the UV absorption by the targeted dioxin, presence of catalytic nTiO2 on soil surface, solvent evaporation rate, as well as vertical gradients of solvents added into the soil columns. Results of dioxin analysis for the soil samples collected at the end of every experiment condition show that the photodegradation enhanced by the nTiO2 presence on the soil surface considerably increased the dioxin removal. Higher removal efficiencies were found for treatments with 15%wt of nTiO2 mixed in the 2-mm surface soil as compared to the 5%wt nTiO2 treatments. The highest removal efficiency (79.6%) was for the sequential intermittent-continuous UV-exposure experiment with nTiO2. Dechlorinated products of 2,3,7,8-TCDD were generally not detected which suggests degradation of targeted dioxin by C-Cl cleavage was negligible. Further modifications to improve removal efficiencies were proposed. Large-scale engineered systems may employ this integrated treatment approach which can also incorporate the reuse of the top soil containing nTiO2 and solvent vapours. With the utilization of natural sunlight such systems would be promisingly suitable for tropical conditions.
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Tetradecane, olefin free, ≥99.0% (GC)
Sigma-Aldrich
Tetradecane, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Supelco
Tetradecane, analytical standard
Sigma-Aldrich
Olive oil, tested according to Ph. Eur.
Supelco
Olive oil, analytical standard
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%