Merck
  • Neurocognitive evidence for revision of treatment targets and guidelines for phenylketonuria.
Neurocognitive evidence for revision of treatment targets and guidelines for phenylketonuria.
The Journal of pediatrics (2014-02-04)
Rianne Jahja, Stephan C J Huijbregts, Leo M J de Sonneville, Jaap J van der Meere, Francjan J van Spronsen
ABSTRACT
To compare the neurocognitive outcomes of patients with phenylketonuria (PKU) to determine whether decreasing phenylalanine (Phe) levels to <240 is preferable to the use of 360 μmol/L as an upper-target Phe level. An additional aim was to establish the influence of biochemical indices other than Phe on neurocognitive outcomes. Patients with PKU (n = 63; mean age 10.8 ± 2.3 years) and healthy controls (n = 73; mean age 10.9 ± 2.2 years) performed computerized tasks measuring neurocognitive functions (inhibitory control, cognitive flexibility, and motor control). Lifetime and concurrent blood Phe levels, Phe-to-tyrosine ratio (Phe:Tyr), and Phe variations were examined in relation to neurocognitive outcomes using nonparametric tests and regression analyses. Patients with PKU with Phe levels ≤240 μmol/L and healthy controls performed equally well. Patients with Phe levels between 240 and 360 μmol/L and ≥360 μmol/L performed more poorly than did controls across tasks. Patients with Phe levels ≤240 μmol/L performed significantly better than patients with levels between 240 and 360 μmol/L on tasks measuring inhibitory control and cognitive flexibility. Absolute Phe levels and Phe variation were the best predictors of motor control, whereas Phe:Tyr were the best predictors of inhibitory control. The results of this study suggest that upper Phe targets should be lowered to optimize neurocognitive outcomes. Moreover, Phe variation and Phe:Tyr appear to be of additional value in treatment monitoring.
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Tyrosine, FG
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
USP
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Phenylalanine, European Pharmacopoeia (EP) Reference Standard
Tyrosine, European Pharmacopoeia (EP) Reference Standard