Merck
  • Mathematical model-based optimization of physico-enzymatic hydrolysis of Pinus roxburghii needles for the production of reducing sugars.
Mathematical model-based optimization of physico-enzymatic hydrolysis of Pinus roxburghii needles for the production of reducing sugars.
Indian journal of experimental biology (2014-01-15)
Siddharth Vats, Devendra Prasad Maurya, Ayushi Jain, Varija Mall, Sangeeta Negi
ABSTRACT
The objective of this study was to optimize the physico-enzymatic pretreatment of P. roxburghii fallen foliage (needles) to produce reducing sugars through response surface methodology (RSM) with central composite face centered design (CCD). Under this, five parameters, i.e., concentration of laccase, cellulose and xylanase, steam explosion pressure and incubation period, at three levels with twenty six runs were taken into account. Cellulase, xylanase and laccase enzymes with activity 4.563, 38.32 and 0.05 IU/mL, respectively, were produced from locally isolated microbial strains. The analysis of variance (ANOVA) was applied for the validation of the predicted model at 95% of confidence level. This model predicted 334 mg/g release of reducing sugars on treating P. roxburghii fallen foliage with 1.18 mL of cellulose, 0.31 mL of xylanase and 0.01 mL of laccase, 14.39 psi steam explosion pressure and 24 h of incubation time. The experimental results obtained were in good agreement to predicted values, making it a reliable optimized model for five factors in combination to predict reducing sugar yield for ethanol production for bio-fuel industry.
Sigma-Aldrich
Laccase from Trametes versicolor, powder, light brown, ≥0.5 U/mg
Sigma-Aldrich
Laccase from Agaricus bisporus, powder, deep brown, ≥4 U/mg
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
Sigma-Aldrich
Cellulase from Aspergillus sp., aqueous solution
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Xylanase from Trichoderma viride, lyophilized powder, 100-300 units/mg protein