Merck
  • Comparison of the shaping ability of GT® Series X, Twisted Files and AlphaKite rotary nickel-titanium systems in simulated canals.
Comparison of the shaping ability of GT® Series X, Twisted Files and AlphaKite rotary nickel-titanium systems in simulated canals.
BMC oral health (2013-12-18)
Raidan Ba-Hattab, Anne-Kathrin Pröhl, Hermann Lang, Dieter Pahncke
ABSTRACT
Efforts to improve the performance of rotary NiTi instruments by enhancing the properties of NiTi alloy, or their manufacturing processes rather than changes in instrument geometries have been reported. The aim of this study was to compare in-vitro the shaping ability of three different rotary nickel-titanium instruments produced by different manufacturing methods. Thirty simulated root canals with a curvature of 35˚ in resin blocks were prepared with three different rotary NiTi systems: AK- AlphaKite (Gebr. Brasseler, Germany), GTX- GT® Series X (Dentsply, Germany) and TF- Twisted Files (SybronEndo, USA).The canals were prepared according to the manufacturers' instructions. Pre- and post-instrumentation images were recorded and assessment of canal curvature modifications was carried out with an image analysis program (GSA, Germany).The preparation time and incidence of procedural errors were recorded. Instruments were evaluated under a microscope with 15 × magnifications (Carl Zeiss OPMI Pro Ergo, Germany) for signs of deformation. The Data were statistically analyzed using SPSS (Wilcoxon and Mann-Whitney U-tests, at a confidence interval of 95%). Less canal transportation was produced by TF apically, although the difference among the groups was not statistically significant. GTX removed the greatest amount of resin from the middle and coronal parts of the canal and the difference among the groups was statistically significant (p < 0.05). The shortest preparation time was registered with TF (444 s) and the longest with GTX (714 s), the difference among the groups was statistically significant (p < 0.05). During the preparation of the canals no instrument fractured. Eleven instruments of TF and one of AK were deformed. Under the conditions of this study, all rotary NiTi instruments maintained the working length and prepared a well-shaped root canal. The least canal transportation was produced by AK. GTX displayed the greatest cutting efficiency. TF prepared the canals faster than the other two systems.
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, 5-10 mm, ≥99.99% trace metals basis (purity exclusive of Na and K content)
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Titanium, IRMM®, certified reference material, 0.5 mm wire
Titanium, IRMM®, certified reference material, 0.5 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis