Merck
Field emission properties of a graphene/polymer composite.
Journal of nanoscience and nanotechnology (2013-11-20)
Shashikant P Patole, Jong Hak Lee, Jae Hong Park, Seong Man Yu, V G Makotchenko, A S Nazarov, V E Fedorov, Dong Wook Shin, Prashant S Alegaonkar, Mahendra A More, Ji-Beom Yoo
ABSTRACT
Thin graphene/polymer sheet composites were fabricated using easily soluble expanded graphite (ESEG), and their field emission (FE) parameters were examined. Due to the high dispersability of ESEG, a stable graphene suspension was prepared by ultrasonication in toluene without the need for a surfactant. The suspension consisted of exfoliated graphene sheets with a thickness of 1 - 2 nm. Using a calendering process, the solution was further shear mixed with ethyl cellulose to obtain a well-dispersed graphene/polymer composite. The composite was screen printed onto a conducing substrate to fabricate the FE cathode layers. The FE measurements were taken in a diode configuration at an applied electrostatic field and inter-electrode distance of 1.7 to 6 V/microm and approximately 200 microm, respectively. The threshold turn-on-field was approximately 3.5 V/microm at a current density of approximately 10 microA/cm2 with a corresponding mean field enhancement factor of 1350 +/- 50. Emission occurred mainly from the edges and bends of the graphene layers. The luminescence uniformity of the composite cathode layers was examined using a phosphor-coated anode.
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Cellulose, acid washed, powder, for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline