Skip to Content
Merck
All Photos(1)

Documents

496537

Sigma-Aldrich

Graphite

rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

Assay

99.995% trace metals basis

form

rod

mol wt

Mw 12.011 g/mol

composition

C

L

150 mm

diam.

3 mm

mp

3652-3697 °C (lit.)

density

2.26 g/cm3 (lit.)

application(s)

battery manufacturing

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Curcumin loaded β-cyclodextrin-magnetic graphene oxide nanoparticles decorated with folic acid receptors as a new theranostic agent to improve prostate cancer treatment.: This study highlights the role of graphite derivatives in enhancing the delivery of therapeutic agents, specifically curcumin, to prostate cancer cells. The use of graphite-based nanoparticles improves targeted drug delivery, a critical innovation in medical device technology and pharmaceutical research (Einafshar et al., 2024).
  • Optimised graphite/carbon black loading of recycled PLA for the production of low-cost conductive filament and its application to the detection of β-estradiol in environmental samples.: Showcases graphite′s application in environmental biosensing technologies, specifically for the detection of contaminants, highlighting its importance in academic and industrial research sectors (Augusto et al., 2024).

Quantity

43.5g = 25 rods

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Craig E Banks et al.
The Analyst, 131(1), 15-21 (2006-01-24)
Different types of carbon based electrodes have emerged over the last few years, significantly changing the scope and sensitivity of electro-analytical methods for the measurement of diverse targets from metal ions through gases to biological markers. This Highlight article shows
Anja Schinwald et al.
Nanotoxicology, 8(8), 824-832 (2013-08-09)
Two-dimensional graphitic carbon, graphene, is a new form of nanomaterial with great potential in a wide variety of applications. It is therefore crucial to investigate the behaviour of graphene in biological systems to assess potential adverse effects that might follow
M A Pimenta et al.
Physical chemistry chemical physics : PCCP, 9(11), 1276-1291 (2007-03-10)
Raman spectroscopy has historically played an important role in the structural characterization of graphitic materials, in particular providing valuable information about defects, stacking of the graphene layers and the finite sizes of the crystallites parallel and perpendicular to the hexagonal
Stephanie Reich et al.
Philosophical transactions. Series A, Mathematical, physical, and engineering sciences, 362(1824), 2271-2288 (2004-10-16)
We present a review of the Raman spectra of graphite from an experimental and theoretical point of view. The disorder-induced Raman bands in this material have been a puzzling Raman problem for almost 30 years. Double-resonant Raman scattering explains their
Craig E Banks et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 21(11), 1263-1268 (2005-12-02)
The recent development, behavior and scope of edge plane pyrolytic graphite electrodes in electroanalysis are overviewed. Similarities to, and advantages, over multi-walled CNT modified electrodes are noted and the wide scope of applications, ranging through gas sensing, stripping voltammetry and

Articles

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service