Merck
  • Preparation and application of graphene-poly (diallyldimethylammoniumchloride)-iron oxide nanoparticles buckypaper for hydrogen peroxide detection.
Preparation and application of graphene-poly (diallyldimethylammoniumchloride)-iron oxide nanoparticles buckypaper for hydrogen peroxide detection.
Journal of nanoscience and nanotechnology (2013-11-20)
Xueque You, Jeejung Kim, Youngmi Kim Pak, James Jungho Pak
ABSTRACT
We have reported the preparation and characterization of a novel, freestanding, paper-like graphene (G)-poly(diallyldimethylammoniumchloride) (PDDA)-Fe3O4 nanoparticles (NPs) composite. This G-based flexible buckypaper (BP) composed of stacked G-PDDA-NP platelets exhibited excellent mechanical properties, superior electrical properties, and enzyme mimetic activity, making it potentially suitable for in electrochemical sensor applications. The negatively charged NPs were immobilized on positively charged G-PDDA through the electrostatic interaction to form nanoscale G-PDDA-NP platelets, which were further assembled by flow-directed assembly to form BP. The resulting BP has macroscopic flexibility and stiffness due to the van der Waals forces between nanoscale G-PDDA-NP platelets and interlocking-tile arrangement of the platelets. The morphology and structure of the individual G-PDDA-NP platelets and the resulting BP were analyzed by using AFM, SEM and EDX. The BP was attached to an Au or Pt electrode to construct a non-enzyme H2O2 chemical sensor. The NPs acted as a "spacer" to increase the distance between the G sheets and decrease the chances of formation of a stacked graphitic structure, thereby increasing the surface area of the G electrode. The Fe3O4 NPs immobilized and embedded in the BP have intrinsic enzyme mimetic activity like natural peroxidase. The high surface area and excellent electrical conductivity of G improved the catalytic properties of NPs. The obtained H2O2, chemical sensor exhibited prominent electrocatalytic activity towards H2O2, with a wide linear range from 10 ppm (approximately 0.3 mM) to 800 ppm (approximately 23 mM), correlation coefficient of 0.986, and a high sensitivity of 218 microA mM(-1) x cm(-2). Such low-cost G-PDDA-NP composite BPs prepared by facile methods pave way towards novel sensors with better performance.
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, 20 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 400,000-500,000 (high molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw <100,000 (very low molecular weight), 35 wt. % in H2O
Sigma-Aldrich
Poly(diallyldimethylammonium chloride) solution, average Mw 200,000-350,000 (medium molecular weight), 20 wt. % in H2O
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O