Merck
  • In vitro evaluation of osteoconductivity and cellular response of zirconia and alumina based ceramics.
In vitro evaluation of osteoconductivity and cellular response of zirconia and alumina based ceramics.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
Ajoy Kumar Pandey, Falguni Pati, Debika Mandal, Santanu Dhara, Koushik Biswas
ABSTRACT
Developed ceria/yttria stabilized zirconia and ceria/yttria stabilized zirconia toughened alumina supported formation of apatite layer when immersed in simulated body fluid without any prior surface treatment. The formed mineral layer was confirmed as hydroxyapatite through X-ray diffraction patterns. The calcium/phosphate atomic ratio obtained from energy dispersive X-ray spectroscopy was found to be little less (Ca/P=1.5) than that of pure hydroxyapatite (Ca/P=1.7) which indicates the probability of mixed type calcium-phosphate compound formation. The achieved thickness of apatite layer was estimated through a surface profilometer and as high as ~17 μm thickness was found after 28 days of soaking. The biocompatibility of the developed materials was ensured through in vitro human osteoblast like cell (MG63) culture on ceramic discs. The morphology of attached cells was characterized through scanning electron microscopy and fluorescent microscopy which show multilayered interconnected cell growth within 8 days of culture period. Moreover, differentiation of MG63 cells was evaluated through MTT assay, total protein content and alkaline phosphatase activity.
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Cerium(IV) oxide, dispersion, nanoparticles, <25 nm particle size, 10 wt. % in H2O
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Cerium(IV) oxide, powder, 99.995% trace metals basis
Sigma-Aldrich
Cerium, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Cerium, chips
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Cerium(IV) oxide, ≥99.0%
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <25 nm particle size (BET)
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Cerium(IV) oxide, dispersion, 20 wt. % colloidal dispersion in 2.5% acetic acid, 30-50 nm avg. part. size
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri